Learn More
Power Blotter Welcome Pack, PVDF
Description
The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter Welcome Pack, PVDF, includes:
- Power Blotter Station
- Power Blotter Cassette
- Power Blotter Select Transfer Stacks, PVDF, regular size
- Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
- Power Blotter 1-Step Transfer Buffer
Features of the Power Blotter System include:
- Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
- Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
- Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
- Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
- Flexible—simultaneous transfer of 1 or 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
- Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Specifications
Specifications
| For Use With (Equipment) | Acrylamide Gels |
| Quantity | 1 Kit |
| Material | PVDF |
| Content And Storage | • Power Blotter Station • Power Blotter Cassette • Power Blotter Select Transfer Stacks, PVDF, regular-size • Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size • Power Blotter 1-Step Transfer Buffer |
| Shipping Condition | Room Temperature |
| Mode of Transfer | Semi-Dry |
| Product Line | Power Blotter |