Learn More
Brady™ PowerMark™ Polyester Labeling Tape
PowerMark™ Polyester Labeling Tape for labelling stainless steel and smooth surfaces. Applies best to very smooth surfaces and ideal for mounting to rigid panels. B-569 adheres to smooth surfaces with the same durability as B-595.
Brand:  Brady™ 013574
 View more versions of this product
Product Code. 11502675
  • 307.00 EUR / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Description
  • For labelling stainless steel and smooth surfaces.
  • Applies best to very smooth surfaces and ideal for mounting to rigid panels.
  • B-569 adheres to smooth surfaces with the same durability as B-595.
Specifications
Specifications
B85-178x15M-569-YL
Permanent Acrylic, Pressure-sensitive
Polyester
Labels
BBP85, PowerMark Industrial Sign and Label Printer
1
1 Roll/Box
MarkWare
-40°C to +100°C
0.003 in.
15.24 m x 177.80 mm
15.24 m
177.8 mm
BBP85 Tape - B-569 178 mm Yellow
Yellow
Thermal Transfer
Facility Identification, Pipe and valve marking
80 Gardner Units
-40°C
1 Ea.
-40°F to +212°F
45 lb./in.
0.076 mm
50 ft.
7 in.