Learn More
Hirschmann™ Polypropylene Powder Funnels
Product Code. 10620512
Click to view available options
Diameter (Metric)Top:
60mm
80mm
100mm
120mm
150mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 10620512
Brand: Hirschmann™ 7760105
Only null left
Add to basket
Add to basket
Ideal for use with powders and other large particles
  • Transparent
Specifications
Diameter (Metric)Top 150mm
Material Polypropylene
Height (Metric) 138 mm
Length (Metric) Stem 22mm
Quantity 5/Pk.