missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PP Reagent Bottles

Product Code. 10653262
Click to view available options
Capacity (Metric):
5000 mL
Diameter (Metric):
178 mm
Height (Metric):
315 mm
Material:
Polypropylene
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from polypropylene. Vitlab™ PP Reagent Bottles offer good chemical resistance.

  • Transparent
  • With screw cap made of polypropylene
  • Good chemical resistance, ideal for long-term storage of liquids Autoclavable at 121 °C (2 bar) according to DIN EN 285
  • Wider mouth for simple filling
  • Ideal for powders and paste-like materials

Specifications

Capacity (Metric) 5000 mL
Material Polypropylene
No. per Case 1Each
No. per Pack 1Each
Packaging 1Each
Quantity 1Each
Height (Metric) 315 mm
Product Type Narrow Mouth Bottle
Diameter (Metric) 178 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'