missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R5E Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23885-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily.Specifications
PPP2R5E | |
Polyclonal | |
Unconjugated | |
PPP2R5E | |
PPP2R5E | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5529 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16537 | |
PPP2R5E | |
PPP2R5E Fusion Protein Ag20891 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |