Learn More
Zwiebel™ Precision E2 Weight
Boxed weight
Brand: Zwiebel™ PBI56824
This item is not returnable.
View return policy
Description
- Precision calibration weight
Specifications
Weight | |
E2 | |
2000 g | |
Balance calibration |