Learn More
Invitrogen™ Primer 5' PFB
Custom Primer
Brand:  Invitrogen™ 11766011
300.75 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10174353
  • 401.00 EUR / Each
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' PFB Primer, 25 nmol
5' PFB Primer
5' Labeled
25 nmol