Learn More
Epredia™ PrintMate™ AS Cassette Printer Accessories
Product Code. 12963815 Shop All Epredia Products
Click to view available options
Product Type:
Cassette Hopper
Collection System
Collection Tray
Computer
Hot Foil Tape
Unit Size:
Each
Pack of 4
Pack of 5
Product Code. 12963815
Brand: Epredia™ B81300033
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Computer
    Description 15 in. All-in-one PC
    For Use With (Equipment) PrintMate AS Cassette Printer, SlideMate Slide Printer