Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human HGF Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-12069-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to human HGF
- human HGF Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human HGF monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
Serum, Plasma: 25 μL; CCS: 50 μL |
Human | |
Cell Culture Supernatant, Plasma, Serum | |
Hepatocyte Growth Factor, Hepapoietin A, Scatter Factor |