Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human IL-27 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX01A-12085-901
Product Code. 15522227
  • 306.00 EUR / 96 tests
Explore more special offers
Includes:
  • concentrated Magnetic Beads coated with monoclonal antibody to human IL-27
  • human IL-27 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human IL-27 monoclonal antibody
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Serum, Plasma: 25 μL; CCS: 50 μL
Human
Cell Culture Supernatant, Plasma, Serum
Interleukin-27, IL27p28