Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human IL-37 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX010-12268-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to human IL-37
- human IL-37 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human IL-37 monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
Interleukin-37, IL-1H4 |
Human | |
IL-37 (IL-1F7) | |
Serum, Plasma: 25 μL; CCS: 50 μL |