Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human IL-37 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX010-12268-901
Product Code. 15582127
  • 306.00 EUR / 96 tests
Estimated Shipment: 01-10-2024
to see stock.
Explore more special offers
Add to basket
Includes:
  • concentrated Magnetic Beads coated with monoclonal antibody to human IL-37
  • human IL-37 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human IL-37 monoclonal antibody
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Cell Culture Supernatant, Plasma, Serum
Interleukin-37, IL-1H4
Human
IL-37 (IL-1F7)
Serum, Plasma: 25 μL; CCS: 50 μL