Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TIMP-1 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-12018-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to human TIMP-1
- human TIMP-1 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human TIMP-1 monoclonal antibody
- Universal Assay Buffer 10x
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- It is recommended to dilute TIMP-1 samples when measured in serum or plasma
- The recommended sample dilution factor is 1:20
Specifications
96 Tests | |
Multiplex Immunoassay | |
Serum, Plasma: 25 μL; CCS: 50 μL |
Human | |
Cell Culture Supernatant, Plasma, Serum | |
Tissue inhibitor of metalloproteinases, EPA |