Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TNF Receptor Superfamily Panel (12 plex)
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX120-15802-901
Includes:
- Human Standard Mix F
- Human Standard Mix L
- Human TNF Receptor Superfamily Bead Mix
- Human TNF Receptor Superfamily Detection Antibody Mix
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit cannot be combined with other ProcartaPlex Simplex or Multiplex Kits
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
Convenience Panel |
Human | |
CD120a (TNF Receptor I), CD120b (TNF Receptor II), CD137 (4-1BB), CD27, CD270 (HVEM), CD30, CD357 (GITR), CD358 (DR6), CD40, CD95 (APO-1/Fas), TRAIL-R1, TRAIL-R2 | |
Serum, Plasma: 25 μL; CCS: 50 μL |