Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TNF Receptor Superfamily Panel (12 plex)
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX120-15802-901
Product Code. 15562477
  • 2990.00 EUR / 96 tests
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Includes:
  • Human Standard Mix F
  • Human Standard Mix L
  • Human TNF Receptor Superfamily Bead Mix
  • Human TNF Receptor Superfamily Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit cannot be combined with other ProcartaPlex Simplex or Multiplex Kits
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Cell Culture Supernatant, Plasma, Serum
Convenience Panel
Human
CD120a (TNF Receptor I), CD120b (TNF Receptor II), CD137 (4-1BB), CD27, CD270 (HVEM), CD30, CD357 (GITR), CD358 (DR6), CD40, CD95 (APO-1/Fas), TRAIL-R1, TRAIL-R2
Serum, Plasma: 25 μL; CCS: 50 μL