Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TNF-RI Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX01A-10203-901
Product Code. 15572137
  • 306.00 EUR / 96 tests
Explore more special offers
Includes:
  • concentrated Magnetic Beads coated with monoclonal antibody to human TNF-RI
  • human TNF-RI Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human TNF-RI monoclonal antibody
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Cell Culture Supernatant, Plasma, Serum
CD120a, TNFR60, TNF Receptor superfamily member 1A
Human
CD120a (TNF Receptor I)
Serum, Plasma: 25 μL; CCS: 50 μL