Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse BTC Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-26042-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to mouse BTC
- mouse BTC Standard (lyophilized)
- mouse Biotin-Conjugate (50x) anti-mouse BTC monoclonal antibody
Description
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
96 Tests | |
Multiplex Immunoassay | |
Cell Culture Supernatant, Plasma, Serum | |
Betacellulin |
Mouse | |
BTC ( Betacellulin) | |
Serum, Plasma: 25 μL; CCS: 50 μL |