missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile PNHS Medical Gloves

Catalog Number 12745865
Click to view available options
Size:
7
10
This item is not returnable. View return policy
This item is not returnable. View return policy
The hypoallergenic accelerator-free and latex-free Profile PNHS™ is the ideal choice for those with sensitive skin. The sterile, extra length, nitrile surgical glove is anatomically shaped to enhance comfort and avoid wearer fatigue. The PNHS offers excellent chemical protection and has good puncture resistance. Measuring 300mm/12” the PNHS provides extra coverage than standard surgical gloves and features a beaded cuff to ensure stability on the arm.
Back Material Nitrile
Certifications/Compliance ISO 9001:2008 & 13485:2003 and Directive 93/42/EEC Annex V
Classification Class IIa Medical Device
Cuff Material Nitrile
Color White
Hand Specific
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Gloves
Size 7
Sterility Sterile
Glove Cuff Style Beaded/Rolled
Packaging Type Boxed
Quantity 50 Pairs
Resistant To Chemicals
Length (Metric) 305 mm
Weight (Metric) 1
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'