Learn More
Heidolph Instruments Guard Hood
Protective cover
Brand:  Heidolph Instruments 569-00010-00
Product Code. 10772757
  • 419.00 EUR / Each
Estimated Shipment: 12-02-2025
Description
  • Strong and transparent guard hood to protect you from implosions
  • Provides easy and immediate access to flask and bath
  • Application range up to 130°C
Order Info
dimType
Specifications
Specifications
Polycarbonate
Protective Cover
1 Ea.
115/230 VAC