Learn More
Eppendorf™ Protein LoBind™ Deepwell Plates
BIOPHARMAGREENER_CHOICE
Product Code. p-7225095 Shop All Eppendorf Products
Click to view available options
Color:
White
White, Clear Bottom
Yellow
No. of Wells:
96
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 8 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
Unit Size:
Pack of 120
Pack of 20
Pack of 40
Pack of 80
Product Code. 10574073
Brand: Eppendorf™ 0030504119
Only null left
Add to basket
Add to basket
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
TRUSTED_SUSTAINABILITY
Specifications
Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 8 Plates
Color Yellow
Product Type Protein LoBind™ Plate
Material Polypropylene
Volume (Metric) Well 500 μL
No. of Wells 96
Sterility Non-sterile
Dnase Free Yes
For Use With (Application) Preparation or Storage of Protein, Peptide or Antibody Samples
Max. Heat Stability 121°C
Max. RCF 6000 x g
Profile Deep well plates, 96 Wells
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 40 Pk.
Well Color Colorless
Dimensions (L x W x H) 85.5 x 127.8 x 27.1 mm