Learn More
Masterflex™ PTFE 3-Way Direct Lift Solenoid Valve
Click to view available options
Voltage:
12 VDC
24 VDC
Maximum Pressure:
18 psi
30 psi
60 psi
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Ideal for use with high-purity or corrosive fluids |
| Max. Operating Temperature (Metric) | 70°C |
| Material | PTFE |
| Quantity | 1 Each |
| Type | 3-way Direct Lift Solenoid Valve |
| Voltage | 24 VDC |
| Maximum Pressure | 60 psi |