Learn More
Fisherbrand™ Conical Joint Clamps
Provides the safest solution for clamping joints. Fisherbrand™ Conical Joint Clamps are manufactured from pure PTFE plastic with totally encapsulated steel springs.
Brand:  Fisherbrand™ 030.040.1
Product Code. 10600313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Adjustable by hand
  • Made to fit standard glass joints
  • Chemically inert and unaffected by lab chemicals
  • Temperature resistant up to 250°C (480°F)
Specifications
Specifications
Conical
PTFE Joint Clamp
Standard glass joints, size 40
2/Pk.
PTFE-encapsulated Steel Spring
White