Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 50
Description
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
- Made of PTFE with thread and 3-part valve locking device
- For use with polished DURAN™ one-way taps
- Grease-free working
Specifications
Specifications
| Plug Bore | 10 mm |
| Material | PTFE |
| Autoclavable | Not Autoclavable |
| Closure Size | 29.2 NS |
| No. per Pack | 10/Pk. |
| For Use With (Application) | For use with polished DURAN™ one-way taps |