Learn More
BRAND™ Straight-Bore PTFE Stopcocks
Product Code. 10374991 Shop All Brand Products
Click to view available options
Fits Tubing ID:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
Unit Size:
Each
Product Code. 10374991
Brand: BRAND™ 88610
Only null left
Add to basket
Add to basket
PTFE construction with polypropylene valve lever. Provides outstanding chemical resistance. Includes integrated hose nozzles.
  • PTFE construction
  • Polypropylene valve
  • With integrated one-way hose nozzle
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 2 mm
Material PTFE
Fits Tubing ID 3 mm
Includes Integrated hose nozzle
Plug Material Polypropylene