Learn More
BRAND™ PTFE Stirring Rod
Shop All Brand Products
Click to view available options
Length (Metric):
200 mm
250 mm
300 mm
Unit Size:
Each
Description
- PTFE construction
- With spatulate ends
Specifications
Specifications
| Includes | Spatulate ends |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 250 mm |
| Material | PTFE |
| Quantity | 1 |