Learn More
Bohlender™ PTFE U-Shaped Stirrer Shafts
Brand:  Bohlender™ C384-17
 View more versions of this product
Product Code. 10257451
  • 275.00 EUR / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Specifications
10 mm
450 mm
U-shaped Stirrer Shaft
Strong, tangential flow with high shear rate in the margin area, little sediments on the wall of the vessel
1 Each