missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPIP51 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
383.00 EUR - 611.00 EUR
Specifications
Antigen | PTPIP51 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331286
|
Bio-Techne
NBP3-17917-25UL |
25 μg |
383.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18362115
|
Bio-Techne
NBP3-17917-100UL |
100 μg |
611.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTPIP51 Polyclonal antibody specifically detects PTPIP51 in Human samples. It is validated for Western BlotSpecifications
PTPIP51 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Cerebral protein 10, FAM82Cmicrotubule-associated protein, family with sequence similarity 82, member A2, FLJ10579, hRMD-3, Protein FAM82A2, Protein FAM82C, Protein tyrosine phosphatase-interacting protein 51, ptpip51, PTPIP51family with sequence similarity 82, member C, regulator of microtubule dynamics 3, regulator of microtubule dynamics protein 3, RMD3, RMD-3, TCPTP-interacting protein 51 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SQRWKRTQRHGRSQSLPNSLDYTQTSDPGRHVMLLRAVPGGAGDASVLPSLPREGQEKVLDRLDFVLTSLVALRREVEELRSSLRGLAGEIVGE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
55177 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title