missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ PU900 Series Medium Duty Polyurethane Gloves

Product Code. 10558645
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides good mechanical resistance to abrasion and cuts. Ansell PU900 Series Medium Duty Polyurethane Gloves provide comfort and dexterity.

  • Mechanical protection gloves
  • Medium duty cut resistance gloves
  • Good abrasion and cut resistance
  • Low lint glove
  • Good comfort and dexterity

Applications:

  • Assembly of white goods
  • Plastics injection and moulding
  • Assembling metal parts and components
  • Aeronautics
  • Finishing works
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Description Gauge: 13
Color White, Gray
Exterior Finish Palm Coated
Material Polyurethane
Product Type Mechanical Protection Gloves
Size 7
Style Construction: Knitted
Glove Cuff Style Knit
Liner Material Spandex Dyneema™
Packaging Type 12 pairs in a bag, 240 pairs in a carton
Series PU900
Length (Metric) 196 to 252 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'