Learn More
Saint Gobain Life Sciences™ Pure Fit™ SIB™ Tri-Clamp
The Pure Fit™ SIB™ Tri-Clamp utilizes the patented BarbLock™ retention and seal system. Pure-Fit™ SIB™ provides a seamless transition that maintains fluid integrity and leak-free connections with complete biocompatibility, for the most demanding applications and stringent test protocols.
Brand:  Saint Gobain Life Sciences™ PFLS375PPAF
 View more versions of this product
Product Code. 17913431
  • 270.00 EUR / Pack of 25
Explore more special offers
Description
Description
  • Eliminates entrapment and leak points that can occur with traditional assembly systems
  • Unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth
  • All materials completely animal-free and BPA-free
  • Polypropylene material is gamma stable
  • USP Class VI and ISO compliant
  • Manufactured and packaged in a certified clean room
  • Available in a variety of sizes
  • Total smooth inner bore (SIB) transition design
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilizes the patented BarbLock™ retention and seal system
  • Compatible with Saint-Gobain Sani-Tech™, C-Flex™ and PharMed™ flexible tubing
  • Applications include: Single-Use Assemblies, Biopharmaceutical Manufacturing, Aseptic and Sterile Fill Lines, Filtration, General Laboratory, Final Fill, Product Sampling, Fluid Transfer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Polypropylene
25 Connectors/Pk.
Tubing Connector
9.5 mm
Biopharmaceutical Manufacturing, Single-use Assembly
Clear