missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Watson-Marlow Tubing PVC Double Segment Manifold Tubing
PVC Double Segment Manifold Tubing for 401U/D1, 401U/DM2 and 401U/DM3 manual/auto control ultra compact pumps.
Brand: Watson-Marlow Tubing 981.0205.000
This item is not returnable.
View return policy
Description
- 401U/D1, 401U/DM2 and 401U/DM3 Manual/auto Control Ultra Compact Pumps
Specifications
0.08 in. | |
PVC | |
0.46 m | |
0.8 mm | |
Double Segment Tubing |
2.05 mm | |
1.5 ft. | |
0.03 in. | |
Purple/Purple |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction