Learn More
Brady™ PVC ISO Safety Signs: Fire Alarm Call Point
Product Code. 15173279
Click to view available options
Style:
Bi-directional
Panoramic
Unit Size:
Each
Product Code. 15173279
Brand: Brady™ 817695
Only null left
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F005-203x203-VPVC-CRD/1
Material PVC
Quantity 1 Ea.
Style Panoramic
For Use With (Application) Indoor & Outdoor
Height (Metric) 203 mm
Thickness 1.591 mm
Width (Metric) 203.00 mm
Temperature Range (Metric) -20 °C-70 °C
Shape Square
Languages Global
Color White on Red