missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55060-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to PYCR2. It has no cross reaction to PYCR1.
PYCR2 belongs to the pyrroline-5-carboxylate reductase family. The function of the PYCR2 protein is not known.Specifications
PYCR2 | |
Polyclonal | |
Unconjugated | |
PYCR2 | |
P5C reductase 2, P5CR 2, P5CR2, PYCR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29920 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96C36 | |
PYCR2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |