Learn More
Pyrex™ Aspirator Bottle with Tubing Side Arm
Product Code. 11317884
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 11317884
Brand: Pyrex 1531/10D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Store and discharge a variety of liquids with this robust aspirator bottle. Pyrex™ Aspirator Bottles with Tubing Side Arm are useful as a delivery or storage container for a wide range of solutions. These bottles feature sturdy ring necks with a heavy duty rims.
•Tubular side arm for attachment of 9mm bore flexible tubing•Unground neck to accept rubber stoppers•Can withstand repeated sterilization (wet or dry)
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Diameter (Metric) Inner Neck 28 mm
Material Borosilicate Glass
Height (Metric) 200 mm
Product Type Aspirator Bottle
Diameter (Metric) 111 mm