Learn More
Pyrex™ Aspirator Bottles, Large
Product Code. p-8338012
Click to view available options
Capacity (Metric):
10,000 mL
20 L
5000 mL
Diameter (Metric) Outer Neck:
100 mm
127 mm
160 mm
Unit Size:
Each
Product Code. 10195034
Brand: Pyrex 1565/16M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for storage of distilled water
  • Wide neck with simple glass cover
  • Supplied with plastic connector and rubber/PTFE sealing ring
  • Accepts either stopcock GP6/10W or glass tubing of 14mm O.D.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5000 mL
Diameter (Metric) Outer Neck 100 mm
For Use With (Application) For storage of distilled water etc
Includes Lid, Plastic connector and Rubber/PTFE sealing ring
Height (Metric) With Closure 320 mm
Material Borosilicate Glass
Mouth Wide
Height (Metric) 320 mm
Product Type Aspirator Bottle
Diameter (Metric) 200 mm