Learn More
Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes
Medium walled test tubes with rimmed edgeManufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
Brand:  Pyrex 1620/20M
 View more versions of this product
Product Code. 11337904
  • 0.39 EUR / Each
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Description
Description
Manufactured from Pyrex™ borosilicate glass to give superior performance over neutral borosilicate and soda lime glass and excellent chemical resistance. Pyrex™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for microbiology culture work.
  • Rimmed medium walls: constructed for optimum performance
  • Robust construction: allows use at high temperatures up to 500deg.C
  • Conforms to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
150 mm
100 Pack
24 mm
55 mL
Borosilicate Glass
Yes
Test Tube