Learn More
Pyrex™ Borosilicate Glass Non-vacuum Desiccators
Product Code. p-7116179
Click to view available options
Height (Metric):
85 mm
120 mm
Diameter (Metric) Outer:
170 mm
220 mm
Unit Size:
Each
Product Code. 12466947
Brand: Pyrex 1590/02D
Only null left
Add to basket
Add to basket
Simple design for drying under atmospheric pressure.
  • Robust desiccator base and cover in borosilicate glass
  • Ground glass flanges
  • Plates are positioned on an internal ledge within the base
  • Choice of 2 sizes - to accommodate 150mm or 200mm diameter plates
  • Accessory metal perforated plate available
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nonvacuum Desiccator
Includes Complete desiccator
Size Plate: 150 mm
Height (Metric) 85 mm
Diameter (Metric) Outer 170 mm