Learn More
Pyrex™ Wide Neck Round Bottom Boiling Flask
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
6 L
missing translation for 'unitSize'
Each
Pack of 10
Pack of 30
Pack of 5
Description
- Complies with ISO 24450 and DIN 12353
- With wide neck to facilitate addition of solid samples etc
- Resistant to chemical corrosion and thermal shock
Specifications
Specifications
| Certifications/Compliance | ISO 24450, DIN 12353 |
| Capacity (Metric) | 4 L |
| Material | Borosilicate Glass |
| Height (Metric) | 315 mm |
| Neck Style | Wide |
| Bottom Shape | Round |
| Diameter (Metric) Outer | 207 mm |
| Closure Size | 67 |