Learn More
Pyrex™ Dessicator Plates
For use with Pyrex vacuum and non-vacuum desiccators
66.15 EUR - 1185.00 EUR
Specifications
Material Metal
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11955879
View Documents
Pyrex
1597/04D
Plate Size: 200mm
66.15 EUR
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
11859932
View Documents
Thermo Scientific
7510B
Plate Size: 150 mm
1185.00 EUR
100mL
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11945879
View Documents
Pyrex
1597/02D
Plate Size: 150 mm
69.35 EUR
Each
Estimated Shipment: 28-04-2025
Description
For use with Pyrex vacuum and non-vacuum desiccators
  • For use with Pyrex vacuum and non-vacuum desiccators
Specifications
Specifications
Metal