Learn More
Pyrex™ Borosilicate Glass Cylinders in Squat Format
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
20 mL
missing translation for 'unitSize'
Pack of 2
Description
- Hexagonal base and squat form delivers stability
- Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
- Heavily beaded rim and spout allow clean and easy dispensing
- Class B
- Complies with ISO 4788
Spécification
Spécification
| Base Type | Hexagonal |
| Scale Color | Blue |
| Shape | Round |
| Type | Graduated Cylinder |
| Tolerance | ±10 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | BS 604, DIN 12680, ISO 4788 |
| Class | B |
| Capacity (Metric) | 1000 mL |
| For Use With (Application) | Confined Spaces |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu