Learn More
Pyrex™ Labware Polyethylene Reservoir Bottle
Feed liquids into a Sofnol burette with easy this filling squeeze bottle. The Pyrex™ labware polyethylene reservoir bottle is a replacement part for the Sofnol, auto zero, picnch clip, class B burette
Brand: Pyrex 3299/06
24.04 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polyethylene
- Squeeze bottle
- For use with Sofnol burette
- Squeeze the bottle to fill the burette
- Release pressure to allow excess liquid to drain back into bottle and set the burrette to zero
- Used for liquid testing
Specifications
1000 mL | |
Sofnol Burette | |
Polyethylene | |
Reservoir Bottle |