Learn More
Pyrex™ Quickfit™ Rotary Evaporator Splash Head with Sinter
Protects the condenser against contamination. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter prevents splash with sinter.
Brand: Quickfit SH16/24
655.50 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Protects the condenser against contamination. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter prevents splash with sinter.
- For use with rotary evaporators
- Prevents possible contamination of the condenser from bumping solvents
- Features a coarse porosity 1 sinter to reduce carry over
- Splash heads feature a 29/32 socket
Specifications
Rotary Evaporator Splash Head with Sinter | |
90mL | |
1/Pk. | |
Cone size: 24/29, Socket size: 29/32 |