Learn More
Corning™ PYREX™ Shorty Disposable Glass Serological Pipets, TD, Sterile, Plugged
Product Code. p-3207927 Shop All Corning Products
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
25 mL
50 mL
Graduations:
0.01 mL
0.1 mL
0.2 mL
0.5 mL
Negative Graduations:
0.5 mL
3 mL
4 mL
5 mL
Packaging Type:
Individually Wrapped
Multi-pack
Unit Size:
Pack of 100
Pack of 200
Pack of 25
Pack of 250
Pack of 400
Pack of 50
Pack of 500
Product Code. 10406653
missing translation for 'mfr': Corning™ 7078B25
Only null left
Add to basket
Add to basket
Ideal for use in confined areas such as hoods
These shorty pipets, calibrated 'To Deliver' with blow-out, are ideal for use in confined areas. The 5, 10, and 25 mL sizes have tooled tops. Negative graduations on each pipet make it applicable for both serological and measuring procedures. Packaged in plastic bags so several can be removed without contaminating the entire pack. ISO color-coding is printed directly on the tear strip.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Cat. No. 7078B-50 features negative and reverse graduations.
Compliance
Calibrated To Deliver (TD) with blow-out.
TRUSTED_SUSTAINABILITY
Specifications
Aspirator Plug Yes
Calibrated Yes
Material Borosilicate Glass
Sterility Sterile
Graduated Yes
Graduations 0.2 mL
Length (English) 11.8 in.
Negative Graduations 5 mL
Tip Style Round
Top Style Tooled
Packaging Type Multi-pack
Volume (English) 0.84 oz.
Color Red
Volume (Metric) 25 mL
Length (Metric) 300 mm
Type Serological Pipet