Learn More
Pyridostigmine-d6 bromide, MedChemExpress
MedChemExpress Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.
Brand: Medchem Express HY-B0207AS
This item is not returnable.
View return policy
Description
Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.Specifications
Pyridostigmine-d6 bromide | |
1 mg | |
Research | |
DMSO : 50 mg/mL (187.16 mM; Need ultrasonic) | |
267.15 | |
98.0% | |
Solid |
2375858-08-3 | |
White | |
C9H7D6BrN2O2 | |
O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])OC1=CC=C[N+](C)=C1.[Br-] | |
267.15 | |
Research |