missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Pyridoxine Control Diet, Modified, Pellet

Diet for rats. Pyridoxine control. Ideal for research use.

779.00 EUR - 2975.00 EUR

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491492
View Documents
MP Biomedicals
0296041810
10 kg
779.00 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
11401502
View Documents
MP Biomedicals
0296041820
20 kg
1370.00 EUR
20kg
Please to purchase this item. Need a web account? Register with us today!
11411502
View Documents
MP Biomedicals
0296041850
50 kg
2975.00 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Diet for rats
  • Pyridoxine control
  • Ideal for research use
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.