Learn More
Axygen™ Qiagen™ Robotic Tips
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
300 μL
1000 μL
1100 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Unit Size:
Pack of 1536
Pack of 2304
Description
Available in volumes of 250, 300, 1000 and 1100 μL.
- Clear and conductive black tips available
- Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips
Specifications
Specifications
| Volume (Metric) | 300 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Black |
| Certifications/Compliance | Non-ANSI |
| Clean Claims | DNase, Rnase Free |
| For Use With (Application) | Non-pyrogenic, liquid level sensing, used for stringent specifications under strict process controls |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | SBS Rack |
| Show More |