Learn More
Grant Instruments™ QR Racks for Grant Heated Circulating Baths
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments QR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
Brand:  Grant Instruments™ QR-LE
 View more versions of this product
Product Code. 11426477
  • 116.60 EUR / Each
Estimated Shipment: 03-01-2025
Description
  • Accommodates tubes as small as 0.5mL or as large as 30mm dia.
  • For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Specifications
44 Tubes
Grant Heated Circulating Baths
0.5 mL Tubes