Learn More
Applied Biosystems™ QuantStudio™ 6 Pro Real-Time PCR System, AB Assurance extended warranty, SmartStart, 96-well, 0.2 mL
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 system
Unit Size:
Each
Description
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- SmartStart Orientation gets users up and running quickly, connects them to their local network, and assists with choosing appropriate reagents/consumables
- 1 year AB Assurance extended warranty including comprehensive service coverage and one planned maintenance visit
- Service plans maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
- During annual maintenance, the FSE will update software if required, clean, lubricate, and align relevant parts and optical systems, functional tests as well as document tasks, results, and findings
Specifications
Specifications
| Block Format | Interchangeable |
| Content And Storage | QuantStudio 6 Pro Real Time PCR System, 96-well, 0.2 mL block, Desktop |
| Product Line | QuantStudio |
| Display Type | 12 in. Color Touchscreen |
| Green Features | Energy Efficient, Fewer Resources used and Less Waste |
| Shipping Condition | Room Temperature |
| Quantity | 1 system |
For Research Use Only