Learn More
SCAT Quick-Lock Connectors
Shop All SCAT Europe Products
Click to view available options
Type:
Quick-Lock Connector (f)
Quick-Lock Connector (m) for 1 Capillary Tube
Quick-Lock Connector (m) for 1 Safety Cap
Quick-Lock Connector Set I
Quick-Lock Connector Set II
Unit Size:
1 set
Each
Description
The Quick-Lock Connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
- Made of chemically resistant glass-filled polypropylene, PEEK™ materials
- Leak protection thanks to integrated shut-off valves
- Closed system for drip-free disconnection with very low air entrapment
- Minimizes contamination and no need to purge air bubbles from the system
- Suitable for capillaries with outer diameter 1.6 (1/16 in.) / 2.3 /3.2 (1/8 in.) mm
- User-friendly, ergonomic design, enables safe connection
- Up to vacuum 15 psi, 1.0 bar pressure resistance
Specifications
Specifications
| For Use With (Equipment) | Capillaries, Tubes and Safety Caps |
| Includes | 1 x PFA-ferrule 1.6/2.3/3.2 mm ID |
| For Use With (Application) | HPLC Systems |
| Type | Quick-Lock Connector (m) for 1 Safety Cap |