Learn More
Quickfit™ Expansion Socket Adapter
Click to view available options
Cone Size:
10/19
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
34/35
40/38
Unit Size:
Each
Pack of 2
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
- Useful for the connection of dissimilar sized ground glass joints
- Large sized socket at the top and small sized cone at the base
Specifications
Specifications
| Cone Size | 14/23 |
| For Use With (Application) | Useful for the connection of dissimilar sized ground Glass joints |
| Material | Borosilicate Glass |
| Product Type | Expansion adapter |
| Standard Taper Joint No. | 29/32 |