Learn More
Quickfit™ Metal Clamps for Flat Flange Joints
Offered in two versions: fork design and spring wire design. Quickfit™ Metal Clamps for Flat Flange Joints are designed to secure corresponding Quickfit™ glassware with flat flange joints.
Brand: Quickfit JC29
This item is not returnable.
View return policy
Description
Offered in two versions: fork design and spring wire design. Quickfit™ Metal Clamps for Flat Flange Joints are designed to secure corresponding Quickfit™ glassware with flat flange joints.
Two distinct styles to meet joint needs
- Fork design: spring loaded to ensure balanced positive pressure to both joint surfaces, finished with corrosion-resistant nickel, suitable for use with spherical joints
- Spring wire design: four touch points for stability, secure swing clamp, stainless steel
Specifications
Fork design | |
Joint Clamp | |
Glassware with flat flange and spherical joints |
5/Pk. | |
Nickel-plated Metal |