Learn More
Contec QuickTask™ Mop Head
QuickTask mop heads that are easily installed onto a hinged mop head frame and handle assembly.
Brand: Contec TASK0800
135.43 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in sterile and non-sterile
- Constructed of Polyester Microfiber
- For use with QuickTask™ Mop
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
019605 | Certificate of Quality | 20/06/2023 | TASK0800 |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%
Product Content Correction
Product Title
Contec QuickTask™ Mop Head > Sterility: Non-sterile
Spot an opportunity for improvement?Share a Content Correction