missing translation for 'onlineSavingsMsg'
Learn More

Quinaldic acid, 98%, Thermo Scientific Chemicals

50.65 EUR - 653.25 EUR

Chemical Identifiers

CAS 93-10-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006752
InChI Key LOAUVZALPPNFOQ-UHFFFAOYSA-N
Synonym quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
PubChem CID 7124
ChEBI CHEBI:18386
IUPAC Name quinoline-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479502
View Documents
Thermo Scientific Alfa Aesar
A10192.06
5 g
50.65 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
11489502
View Documents
Thermo Scientific Alfa Aesar
A10192.14
25 g
177.89 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
11499502
View Documents
Thermo Scientific Alfa Aesar
A10192.22
100 g
653.25 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-10-7
173.17
LOAUVZALPPNFOQ-UHFFFAOYSA-N
7124
quinoline-2-carboxylic acid
C10H7NO2
MFCD00006752
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
CHEBI:18386
OC(=O)C1=CC=C2C=CC=CC2=N1

Specifications

93-10-7
C10H7NO2
126322
quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
OC(=O)C1=CC=C2C=CC=CC2=N1
173.17
CHEBI:18386
98%
156°C to 159°C
MFCD00006752
14,8046
LOAUVZALPPNFOQ-UHFFFAOYSA-N
quinoline-2-carboxylic acid
7124
173.17
Quinaldic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 202-218-3

missing translation for 'rtecsNumber' : UZ9100000

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.